Learn More
Invitrogen™ Lamin B2 Recombinant Rabbit Monoclonal Antibody (1S9M6)

Description
Immunogen sequence: YILRAGQMVT VWAAGAGVAH SPPSTLVWKG QSSWGTGESF RTVLVNADGE EVAMRTVKKS SVMRENENGE EEEEEAEFGE EDLFHQQGDP RTTSRGCYVM.
Specifications
Specifications
| Antigen | Lamin B2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 1S9M6 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | LMNB2 |
| Gene Accession No. | P21619, Q03252 |
| Gene Alias | EPM9; fc15d06; im:7142331; LAMB2; lamin; lamin A/C; lamin B2; lamin B2 S homeolog; lamin B3; lamin-B2; Lamin-L(II); LMN2; LMNA; lmnb2; lmnb2.S; lmnb2-a; lmnb2-b; MGC2721; RGD1563803; unnamed protein product; wu:fb94e05; wu:fb95e12; wu:fc15d06; wu:fc49h03; XELAEV_18009631mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.