Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lamin B2 Monoclonal Antibody (D5-C2)
GREENER_CHOICE

Catalog No. PIMA544617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544617 Supplier Invitrogen™ Supplier No. MA544617
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lamin B2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Nuclear lamins form a network of intermediate-type filaments at the nucleoplasmic site of the nuclear membrane. Two main subtypes of nuclear lamins can be distinguished, i.e. A-type lamins and B-type lamins. The A-type lamins comprise a set of three proteins arising from the same gene by alternative splicing, i.e. lamin A, lamin C and lamin Adel 10, while the B-type lamins include two proteins arising from two distinct genes, i.e. lamin B1 and lamin B2. Recent evidence has revealed that mutations in A-type lamins give rise to a range of rare but dominant genetic disorders, including Emery-Dreifuss muscular dystrophy, dilated cardiomyopathy with conduction-system disease and Dunnigan-type familial partial lipodystrophy. In addition, the expression of A-type lamins coincides with cell differentiation and as A-type lamins specifically interact with chromatin, a role in the regulation of differential gene expression has been suggested for A-type lamins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lamin B2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone D5-C2
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene LMNB2
Gene Accession No. P21619, Q03252
Gene Alias EPM9; fc15d06; im:7142331; LAMB2; lamin; lamin A/C; lamin B2; lamin B2 S homeolog; lamin B3; lamin-B2; Lamin-L(II); LMN2; LMNA; lmnb2; lmnb2.S; lmnb2-a; lmnb2-b; MGC2721; RGD1563803; unnamed protein product; wu:fb94e05; wu:fb95e12; wu:fc15d06; wu:fc49h03; XELAEV_18009631mg
Gene Symbols LMNB2
Host Species Mouse
Immunogen Synthetic peptide within Human LMNB2 aa 571-620/620.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16907, 299625, 30196, 84823
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.