Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lamin B2 Antibody (OTI2C6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272303H
Description
Lamin B2 Monoclonal specifically detects Lamin B2 in Human samples. It is validated for Western Blot.Specifications
Lamin B2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
LMNB2 | |
Full length recombinant protein of human Lamin B2 (NP_116126) produced in HEK293T cells. | |
0.1 mL | |
84823.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2C6 | |
Western Blot | |
lamin B2, lamin-B2, LMN2LAMB2, MGC2721 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction