Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminaran from Eisenia Bicyclis, TCI America™
SDP

Catalog No. p-7131816
Click to view available options
Quantity:
1 g
25 g

Chemical Identifiers

CAS 9008-22-4
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00466918
InChI Key DBTMGCOVALSLOR-DEVYUCJPSA-N
Synonym laminarin, laminaran
PubChem CID 46173707
IUPAC Name (2S,3R,4S,5R,6R)-4-[(2S,3R,4S,5R,6R)-3,5-dihydroxy-6-(hydroxymethyl)-4-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,5-triol
SMILES C(C1C(C(C(C(O1)O)O)OC2C(C(C(C(O2)CO)O)OC3C(C(C(C(O3)CO)O)O)O)O)O)O

Specifications

Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material Laminaran from Eisenia Bicyclis
EINECSNumber (8)-0073
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.