Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin A/B Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA184171

Catalog No. PA184171


Description

Description

PA1-84171 recognizes both A (400 kD) and B (220 kD) chains of laminin. A suggested positive control for immunohistochemical applicationss is human kidney.

Basement membrane components play an important role in cell adhesion, locomotion, differentiation and embryonic compartmentalization. Laminin anchors cells to the basement membrane through interactions with integrins at the cell surface and collagens in the basement membrane.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Laminin A/B
Polyclonal
Unconjugated
LAMA1
AA408497; alternatively spliced laminin gamma2 chain; AW211941; B2T; basement membrane adhesion protein; BM600; BM600-100kDa; C77966; C80098; C81607; Cell-scattering factor 140 kDa subunit; CLM; CSF; CSF 140 kDa subunit; D130003D08Rik; EBR2; EBR2A; Epiligrin subunit gamma; extra-cellular matrix protein; gamma 1 (formerly LAMB2); hypothetical protein; kalinin subunit gamma; Kalinin/nicein/epiligrin 100 kDa subunit; Ladsin 140 kDa subunit; LAMA; Lama1; Lama-1; LAMB1; Lamb-1; Lamb1-1; LAMB2; Lamb-2; LAMB2T; LAMC1; LAMC2; laminin 5 gamma 2 subunit; laminin A chain; laminin alpha 1; laminin alpha 1 chain; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin B1s chain; Laminin B2 chain; laminin B2t chain; laminin beta 2 subunit chain; laminin beta 2-like chain; laminin beta-1-1 chain; Laminin beta-1-2 chain; laminin chain B3; laminin chain beta 2; laminin gamma 1; laminin S; laminin subunit alpha 1; laminin subunit alpha-1; laminin subunit beta 1; laminin subunit beta 2; Laminin subunit beta-1; laminin subunit beta-1 variant; laminin subunit beta-2; laminin subunit gamma 1; laminin subunit gamma 2; laminin subunit gamma-1; Laminin subunit gamma-2; laminin, alpha 1; laminin, beta 1; laminin, beta 2; laminin, beta 2 (laminin S); laminin, gamma 1 (formerly LAMB2); laminin, gamma 2; laminin-1; laminin-1 subunit alpha; Laminin-1 subunit beta; Laminin-1 subunit gamma; Laminin-10 subunit beta; laminin-10 subunit gamma; Laminin-11 subunit beta; laminin-11 subunit gamma; Laminin-12 subunit beta; laminin-14 subunit beta; Laminin-15 subunit beta; laminin-2 subunit beta; Laminin-2 subunit gamma; laminin-3 subunit alpha; Laminin-3 subunit beta; laminin-3 subunit gamma; Laminin-4 subunit beta; laminin-4 subunit gamma; laminin-5 gamma 2; Laminin-5 subunit gamma; Laminin-6 subunit beta; laminin-6 subunit gamma; Laminin-7 subunit beta; laminin-7 subunit gamma; Laminin-8 subunit beta; laminin-8 subunit gamma; Laminin-9 subunit beta; laminin-9 subunit gamma; LAMNB2; Lams; Large adhesive scatter factor 140 kDa subunit; LIS5; Nicein subunit gamma; NPHS5; npht; PTBHS; SLAM; S-LAM alpha; S-LAM beta; S-LAM gamma; S-LAM-alpha; S-laminin subunit alpha; S-laminin subunit beta; S-laminin subunit gamma
Rabbit
50 μL
Primary
Avian, Canine, Chicken, Human, Mouse, Rabbit, Rat
Antibody
IgG
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Conc. Not Determined
whole serum with 0.09% sodium azide
P02469, P07942, P15800, P19137, P25391, P55268, Q01635, Q61292
LAMA1, LAMB1, LAMB2
Purified laminin from Engelbreth-Holm-Swarm (EHS) tumor.
RUO
100009388, 100357482, 100358679, 102156629, 16772, 16777, 16779, 25473, 284217, 298941, 316758, 373980, 374016, 3912, 3913, 396478, 475882, 476626
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.