Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Laminin alpha 1 Antibody (775016), mFluor Violet 500 SE, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4187MFV500
Description
Laminin alpha 1 Monoclonal antibody specifically detects Laminin alpha 1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| Laminin alpha 1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a | 
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 775016 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| LAMA, LAMA1 laminin, alpha 1, S-LAM-alpha | |
| E. coli-derived recombinant human Laminin alpha 1, Leu22-Met269, Accession # P25391 | |
| RUO | |
| 284217 | |
| Store at 4C in the dark. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction