Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin alpha-1 Monoclonal Antibody (CL2968)
GREENER_CHOICE

Catalog No. PIMA531380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531380 Supplier Invitrogen™ Supplier No. MA531380
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: LIVQGRGLID AAAAQTDAVQ DALEHLEDHQ DKLLLWSAKI RHHIDDLVMH MSQRNAVDLV YRAEDHATEF QRLADVLYSG LENIRNVSLN ATSAA Highest antigen sequence indentity to the following orthologs: Mouse - 62%, Rat - 61%.

Laminin antibodies are widely used to label blood vessels and basement membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin alpha-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL2968
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAMA1
Gene Accession No. P25391
Gene Alias LAMA; LAMA1; Laminin A chain; laminin subunit alpha 1; laminin subunit alpha-1; laminin, alpha 1; Laminin-1 subunit alpha; Laminin-3 subunit alpha; PTBHS; S-LAM alpha; S-LAM-alpha; S-laminin subunit alpha
Gene Symbols LAMA1
Host Species Mouse
Immunogen Recombinant protein fragment of Human LAMA1 (Product #RP-108162)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284217
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.