Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin alpha-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119248
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119248 200 μL
PIPA5122092 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119248 Supplier Invitrogen™ Supplier No. PA5119248
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Laminin alpha-1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P), ELISA

Laminin antibodies are widely used to label blood vessels and basement membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin alpha-1
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene LAMA1
Gene Accession No. 0
Gene Alias LAMA; LAMA1; Laminin A chain; laminin subunit alpha 1; laminin subunit alpha-1; laminin, alpha 1; Laminin-1 subunit alpha; Laminin-3 subunit alpha; PTBHS; S-LAM alpha; S-LAM-alpha; S-laminin subunit alpha
Gene Symbols LAMA1
Host Species Rabbit
Immunogen Recombinant protein Laminin Alpha 1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316758
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.