Learn More
Invitrogen™ Laminin alpha-2 Monoclonal Antibody (CL3450)

Description
Immunogen sequence: DKLKPIKELE DNLKKNISEI KELINQARKQ ANSIKVSVSS GGDCIRTYKP EIKKGSYNNI VVNVKTAVAD NLLFYLGSAK FIDFLAIEMR KGKVSFLWDV GSGVGRVEYP DLTIDDSYWY RIVASRTGRN GT Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 98%. Binds to an epitope located within the peptide sequence DKLKPIKELEDNLKK as determined by overlapping synthetic peptides.
Specifications
Specifications
| Antigen | Laminin alpha-2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | CL3450 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Lama2 |
| Gene Accession No. | P24043, Q60675 |
| Gene Alias | 5830440B04; dy; dystrophia muscularis; Lama2; laminin alpha-2; laminin M; Laminin M chain; laminin subunit alpha 2; laminin subunit alpha-2; laminin, alpha 2; laminin-12 subunit alpha; Laminin-2 subunit alpha; laminin-4 subunit alpha; LAMM; mer; merosin; Merosin heavy chain; mKIAA4087 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.