Learn More
Invitrogen™ Laminin alpha-2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA549929
Catalog No. PA549929
Only null left
Add to Cart
Description
Description
The antibody detects endogenous levels of total Laminin alpha-2 protein.
Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Laminin alpha-2
Polyclonal
Unconjugated
Lama2
5830440B04; dy; dystrophia muscularis; Lama2; laminin alpha-2; laminin M; Laminin M chain; laminin subunit alpha 2; laminin subunit alpha-2; laminin, alpha 2; laminin-12 subunit alpha; Laminin-2 subunit alpha; laminin-4 subunit alpha; LAMM; mer; merosin; Merosin heavy chain; mKIAA4087
Rabbit
Antigen affinity chromatography
RUO
16773, 3908
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P24043, Q60675
Lama2
Synthesized peptide derived from internal of human LAMA2.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.