Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin alpha 4 Antibody (OTI5G8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245582
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
Laminin alpha 4 Monoclonal specifically detects Laminin alpha 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Laminin alpha 4 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
DKFZp686D23145, LAMA3LAMA4*-1, laminin alpha 4 chain, laminin subunit alpha-4, laminin, alpha 4, Laminin-14 subunit alpha, Laminin-8 subunit alpha, Laminin-9 subunit alpha | |
Mouse | |
199.8 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
OTI5G8 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
LAMA4 | |
Human recombinant protein fragment corresponding to amino acids 25-270 of human LAMA4(NP_001098676) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
3910 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction