Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin alpha-4 Monoclonal Antibody (CL3183)
GREENER_CHOICE

Catalog No. PIMA524650
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524650 Supplier Invitrogen™ Supplier No. MA524650
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: ENLLNQAREL QAKAESSSDE AVADTSRRVG GALARKSALK TRLSDAVKQL QAAERGDAQQ RLGQSRLITE EANRTTMEVQ QATAPMANNL TNWSQNLQHF DSSAYNTAVN SARDAVRNLT EVVPQLLDQL RTVEQKRPAS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%. Binds to an epitope located within the peptide sequence MANNLTNWSQNLQHF as determined by overlapping synthetic peptides.

Laminin is a non-collagenous protein of basement membranes. By rotary shadowing it has a cross-like shape, consisting of one long arm, (400 kD) and three short arms (200 kD). Antibodies to laminin localize exclusively in basement membranes. Laminin is found throughout the basement membrane, but the reaction with the antibodies to laminin is more intense in the lamina lucida, the part of the basement membrane adjacent to the cell membrane. Laminin binds to type IV collagen, a component exclusively localized in the basement membrane. Laminin has been shown to play a role in cell adhesion and attachment in vivo and in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin alpha-4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL3183
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAMA4
Gene Accession No. P97927, Q16363
Gene Alias CMD1JJ; LAMA3; LAMA4; LAMA4*-1; laminin [a]4; laminin alpha 4 chain; laminin subunit alpha 4; laminin subunit alpha-4; laminin, alpha 4; laminin-14 subunit alpha; Laminin-8 subunit alpha; laminin-9 subunit alpha; RGD1560062
Gene Symbols LAMA4
Host Species Mouse
Immunogen Recombinant protein fragment of Human LAMA4 (Product #RP-90731)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16775, 309816, 3910
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.