Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin alpha-4 Polyclonal Antibody
Laminin alpha-4 Polyclonal Antibody
Supplier: Thermo Scientific PA553365
Description
Laminin alpha-4 Polyclonal Antibody for IHC

Specifications
Laminin alpha-4 | |
Polyclonal | |
Unconjugated | |
LAMA4 | |
CMD1JJ; LAMA3; LAMA4; LAMA4*-1; laminin [a]4; laminin alpha 4 chain; laminin subunit alpha 4; laminin subunit alpha-4; laminin, alpha 4; laminin-14 subunit alpha; Laminin-8 subunit alpha; laminin-9 subunit alpha; RGD1560062 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3910 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q16363 | |
LAMA4 | |
Recombinant protein corresponding to Human Laminin alpha-4. Recombinant protein control fragment (Product #RP-90731). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction