Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin alpha 5 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP172050
Description
Laminin alpha 5 Polyclonal specifically detects Laminin alpha 5 in Human samples. It is validated for Peptide ELISA.Specifications
Laminin alpha 5 | |
Polyclonal | |
Peptide ELISA Detection limit 1:32000 | |
NP_005551.3 | |
LAMA5 | |
Peptide with sequence EACTYRPSAQQSGDN corresponding to internal region according to NP_005551.3. | |
100 μg | |
Primary | |
Human | |
IgG |
ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
KIAA0533, KIAA1907, laminin alpha-5 chain, laminin subunit alpha-5, laminin, alpha 5, Laminin-10 subunit alpha, Laminin-11 subunit alpha, Laminin-15 subunit alpha | |
Goat | |
Affinity Purified | |
RUO | |
3911 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction