Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin alpha-5 Monoclonal Antibody (CL3118)
GREENER_CHOICE

Catalog No. PIMA524649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524649 Supplier Invitrogen™ Supplier No. MA524649
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: LNLSSIILDV NQDRLTQRAI EASNAYSRIL QAVQAAEDAA GQALQQADHT WATVVRQGLV DRAQQLLANS TALEEAMLQE QQRLGLVWAA L Binds to an epitope located within the peptide sequence RQGLVDRAQQ as determined by overlapping synthetic peptides. Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 62%.

Components of the extracellular matrix exert myriad effects on tissues throughout the body. In particular, the laminins, a family of heterotrimeric extracellular glycoproteins, affect tissue development and integrity in such diverse organs as the kidney, lung, skin, and nervous system. It is thought that laminins mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminins function as heterotrimeric complexes of alpha, beta, and gamma chains, with each chain type representing a different subfamily of proteins. The protein encoded by this gene belongs to the alpha subfamily of laminin chains and is a major component of basement membranes. Two transcript variants encoding different isoforms have been found for this gene, but the full-length nature of one of them has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin alpha-5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL3118
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAMA5
Gene Accession No. O15230
Gene Alias [a]5; AA408760; AA408762; AI853660; KIAA0533; KIAA1907; LAMA5; laminin alpha-5 chain; laminin subunit alpha 5; laminin subunit alpha-5; laminin, alpha 5; laminin-10 subunit alpha; Laminin-11 subunit alpha; laminin-15 subunit alpha; laminin-511; mKIAA0533; RP11-157P1.6
Gene Symbols LAMA5
Host Species Mouse
Immunogen Recombinant protein fragment of Human LAMA5 (Product #RP-108178)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3911
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.