Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin alpha 5 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1086R

Description
Components of the extracellular matrix exert myriad effects on tissues throughout the body. In particular, the laminins, a family of heterotrimeric extracellular glycoproteins, affect tissue development and integrity in such diverse organs as the kidney, lung, skin, and nervous system. It is thought that laminins mediate the attachment, migration, and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. Laminins function as heterotrimeric complexes of alpha, beta, and gamma chains, with each chain type representing a different subfamily of proteins. The protein encoded by this gene belongs to the alpha subfamily of laminin chains and is a major component of basement membranes. Two transcript variants encoding different isoforms have been found for this gene, but the full-length nature of one of them has not been determined.Specifications
Laminin alpha 5 | |
Polyclonal | |
Unconjugated | |
LAMA5 | |
[a]5; AA408760; AA408762; AI853660; KIAA0533; KIAA1907; LAMA5; laminin alpha-5 chain; laminin subunit alpha 5; Laminin subunit alpha-5; laminin, alpha 5; Laminin-10 subunit alpha; Laminin-11 subunit alpha; Laminin-15 subunit alpha; laminin-511; mKIAA0533; RP11-157P1.6 | |
Rabbit | |
Protein A | |
RUO | |
3911 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O15230 | |
LAMA5 | |
KLH conjugated synthetic peptide derived from human Laminin alpha 5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction