Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin alpha5 Mouse anti-Human, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1924C25UG
Description
This mouse monoclonal Anti-Laminin alpha5, clone 4C7 (Ascites Free) Immunohistochemistry, ELISA and Neutralizing, for the detection of Laminin alpha 5.
Specifications
Laminin alpha5 | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunoprecipitation, Immunofluorescence, Immunohistochemistry, ELISA, Neutralization | |
Please refer to lot specific datasheet. | |
LAMA5,KIAA0533,KIAA1907 | |
Full length purified human laminin alpha 5. | |
25 μg | |
Cell Structure | |
Clone 4C7 reacts with the globular domain of the alpha5 chain of intact laminin and blocks the neurite stimulating activity of laminin. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction