Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin Antibody, Allophycocyanin/Cy7, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300144APCCY7
Description
Laminin Polyclonal specifically detects Laminin in Human, Mouse, Rat, Chinese Hamster, Invertebrate, Mammal, Rabbit, Sheep samples. It is validated for Flow Cytometry.Specifications
Laminin | |
Polyclonal | |
Flow Cytometry | |
LAMA, Laminin A chain, laminin subunit alpha-1, laminin, alpha 1, Laminin-1 subunit alpha, Laminin-3 subunit alpha, PTBHS, S-LAM alpha, S-laminin subunit alpha | |
Rabbit | |
IgG purified | |
Angiogenesis, Apoptosis, Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors | |
284217.0 | |
Human, Mouse, Rat, Chinese Hamster, Invertebrate, Mammalia, Rabbit, Sheep | |
IgG |
Flow Cytometry | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
LAMA1 | |
Laminin Antibody was made to Laminin 111 isolated from mouse Engelbreth-Holm-Swarm (EHS) sarcoma cells. [UniProt# P19137] | |
0.1 mL | |
Primary | |
Laminin Antibody is pan-specific and reacts well with all Laminin isoforms tested: Laminin-1 (alpha-1, beta-1, and gamma-1) and Laminin-2 (alpha-2, beta-1, and gamma-1). | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction