Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin Antibody (MEC5), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP151759
Description
Laminin Monoclonal specifically detects Laminin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Laminin | |
Monoclonal | |
Unconjugated | |
Tissue culture supernatant with 0.05% Sodium Azide | |
LAMA1 | |
Laminin Antibody (MEC5) was made to the Laminin antigen. | |
Tissue culture supernatant | |
RUO | |
Primary | |
Laminin Antibody (MEC5) is directed against the P1 fragment of laminin. They are species-independent as it has been shown that they bind to laminin of rat, mouse and humans (Aten et al., 1995). MEC5 is an auto-antibody derived from DZB rats that had developed membranous glomerulopathy upon exposure to mercuric chloride (Aten et al. 1995).MEC5 is an auto-antibody derived from DZB rats that had developed membranous glomerulopathy upon exposure to mercuric chloride (Aten et al. 1995). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Immunohistochemistry, Immunohistochemistry (Frozen) | |
MEC5 | |
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
LAMA, Laminin A chain, laminin subunit alpha-1, laminin, alpha 1, Laminin-1 subunit alpha, Laminin-3 subunit alpha, PTBHS, S-LAM alpha, S-laminin subunit alpha | |
Rat | |
337 kDa | |
0.25 mL | |
Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors | |
284217 | |
Human, Mouse, Rat | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction