Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 72 publications

$278.00 - $594.00

Specifications

Antigen Laminin
Concentration 1 mg/ml
Dilution Western Blot 1:100 - 1:5000, Flow Cytometry, Immunohistochemistry 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:500 - 1:2000, Immunohistochemistry-Frozen 1:500 - 1:2000, Immunohistochemistry Free-Floating 1:1000 - 1:5000
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB300144SS
SDP
View Documents
Novus Biologicals
NB300144SS
0.025 mL
Each for $278.00
Only null left
Add to Cart
 
NB300144
SDP
View Documents
Novus Biologicals
NB300144
0.1 mL
Each for $594.00
Only null left
Add to Cart
 
Description

Description

Laminin Polyclonal specifically detects Laminin in Human, Mouse, Rat, Chinese Hamster, Invertebrate, Mammal, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
Specifications

Specifications

Laminin
Western Blot 1:100 - 1:5000, Flow Cytometry, Immunohistochemistry 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:500 - 1:2000, Immunohistochemistry-Frozen 1:500 - 1:2000, Immunohistochemistry Free-Floating 1:1000 - 1:5000
Polyclonal
Purified
RUO
Human, Mouse, Rat, Chinese Hamster, Invertebrate, Mammalia, Rabbit, Sheep
P19137
284217
Laminin Antibody was made to Laminin 111 isolated from mouse Engelbreth-Holm-Swarm (EHS) sarcoma cells. [UniProt# P19137]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
337 kDa
1 mg/ml
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Angiogenesis, Apoptosis, Cancer, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors
50% PBS, 50% Glycerol with 5mM Sodium Azide
LAMA, Laminin A chain, laminin subunit alpha-1, laminin, alpha 1, Laminin-1 subunit alpha, Laminin-3 subunit alpha, PTBHS, S-LAM alpha, S-laminin subunit alpha
LAMA1
IgG
IgG purified
Laminin Antibody is pan-specific and reacts well with all Laminin isoforms tested: Laminin-1 (alpha-1, beta-1, and gamma-1) and Laminin-2 (alpha-2, beta-1, and gamma-1).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.