Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin B2 gamma 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986345
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6345 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6345 Supplier Bioss Supplier No. BS5122R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.

Specifications

Antigen Laminin B2 gamma 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LAMC1
Gene Accession No. P11047, P55268
Gene Alias AA408497; AW211941; B2e; Lama; Lamb2; Lamb-2; LAMC1; Laminin B1s chain; Laminin B2 chain; Laminin chain B3; laminin chain beta 2; laminin S; laminin subunit beta 2; Laminin subunit beta-2; laminin subunit gamma 1; Laminin subunit gamma-1; laminin, beta 2; laminin, beta 2 (laminin S); laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); Laminin-1 subunit gamma; Laminin-10 subunit gamma; Laminin-11 subunit beta; Laminin-11 subunit gamma; Laminin-14 subunit beta; Laminin-15 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit beta; Laminin-3 subunit gamma; Laminin-4 subunit beta; Laminin-4 subunit gamma; Laminin-6 subunit gamma; Laminin-7 subunit beta; Laminin-7 subunit gamma; Laminin-8 subunit gamma; Laminin-9 subunit beta; Laminin-9 subunit gamma; LAMS; LOC100514785; NPHS5; npht; SLAM; S-LAM beta; S-LAM gamma; S-laminin subunit beta; S-laminin subunit gamma
Gene Symbols LAMB2, LAMC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Laminin B2 gamma 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3913, 3915
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.