Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin beta-1 Recombinant Rabbit Monoclonal Antibody (6I9T4)
GREENER_CHOICE

Catalog No. PIMA535495
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535495 Supplier Invitrogen™ Supplier No. MA535495
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: KKTLDGELDE KYKKVENLIA KKTEESADAR RKAEMLQNEA KTLLAQANSK LQLLKDLERK YEDNQRYLED KAQELARLEG EVRSLLKDIS QKVAVYSTCL.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin beta-1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 6I9T4
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LAMB1
Gene Accession No. P02469, P07942
Gene Alias C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015
Gene Symbols LAMB1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16777, 298941, 3912
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.