Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin beta-1 Monoclonal Antibody (CL2970)
GREENER_CHOICE

Catalog No. PIMA535004
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535004 Supplier Invitrogen™ Supplier No. MA535004
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: GPRCDKCTRG YSGVFPDCTP CHQCFALWDV IIAELTNRTH RFLEKAKALK ISGVIGPYRE TVDSVERKVS EIKDILAQSP AAEPLKNIGN LFEEAEKLIK DVTEMMAQVE VKLSDTTSQS NSTAKELDSL QTEAESLDNT VKELAEQLEF Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 89%.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin beta-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL2970
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LAMB1
Gene Accession No. P07942
Gene Alias C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015
Gene Symbols LAMB1
Host Species Mouse
Immunogen Recombinant protein fragment of Human LAMB1 (Product #RP-102390)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3912
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.