Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin beta-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527271
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527271 Supplier Invitrogen™ Supplier No. PA527271
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299, HeLa, NIH-3T3. Predicted reactivity: Mouse (88%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin beta-1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene LAMB1
Gene Accession No. P02469, P07942
Gene Alias C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015
Gene Symbols LAMB1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human Laminin beta 1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16777, 3912
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.