Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Laminin beta-1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582138
Description
Immunogen sequence: NQVVSLSPGS RYVVLPRPVC FEKGTNYTVR LELPQYTSSD SDVESPYTLI DSLVLMPYCK SLDIFTVGGS GDGVVTNSAW ETFQRYRCLE NSRSVVKTPM TDVCRNIIFS ISALLHQTGL ACECDPQGSL SSVCDPNGG.
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
Specifications
Laminin beta-1 | |
Polyclonal | |
Unconjugated | |
LAMB1 | |
C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3912 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P07942 | |
LAMB1 | |
Recombinant protein corresponding to Human LAMB1. Recombinant protein control fragment (Product #RP-102389). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction