Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin beta 1 Rabbit anti-Human, Mouse, Rat, Clone: 6I9T4, Novus Biologicals™

Rabbit Monoclonal Antibody
$206.50 - $492.50
Specifications
Antigen | Laminin beta 1 |
---|---|
Clone | 6I9T4 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Laminin beta 1 Monoclonal antibody specifically detects Laminin beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Laminin beta 1 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3912 | |
IgG | |
Affinity purified |
6I9T4 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CLM, cutis laxa with marfanoid phenotype, Laminin B1 chain, laminin subunit beta-1, laminin, beta 1, Laminin-1 subunit beta, Laminin-10 subunit beta, Laminin-12 subunit beta, Laminin-2 subunit beta, Laminin-6 subunit beta, Laminin-8 subunit beta, MGC142015 | |
A synthetic peptide corresponding to a sequence within amino acids 1687-1786 of human Laminin beta 1 (P07942). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title