Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin beta-2/gamma-1 Monoclonal Antibody (D18), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA511992
Description
MA5-11992 targets Laminin B2/gamma1 in IHC (P) applications and shows reactivity with Guinea Pig, Human, and Rat samples. The MA5-11992 immunogen is rat glomerular basement membrane.
Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
Specifications
Laminin beta-2/gamma-1 | |
Monoclonal | |
0.4 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P11047, P55268 | |
LAMB2, LAMC1 | |
Rat glomerular basement membrane | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Immunohistochemistry (Paraffin) | |
D18 | |
Unconjugated | |
LAMC1 | |
AA408497; AW211941; B2e; basement membrane protein; Lama; Lamb2; Lamb-2; Lamc1; Laminin B1s chain; laminin B2 chain; laminin beta 2; laminin beta 2 chain; laminin chain B3; laminin chain beta 2; laminin gamma 1 chain; Laminin S; laminin subunit beta 2; laminin subunit beta-2; laminin subunit gamma 1; laminin subunit gamma-1; laminin, beta 2; laminin, beta 2 (laminin S); laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1 subunit gamma; Laminin-10 subunit gamma; laminin-11 subunit beta; Laminin-11 subunit gamma; laminin-14 subunit beta; Laminin-15 subunit beta; laminin-2 subunit gamma; Laminin-3 subunit beta; laminin-3 subunit gamma; Laminin-4 subunit beta; laminin-4 subunit gamma; Laminin-6 subunit gamma; laminin-7 subunit beta; Laminin-7 subunit gamma; laminin-8 subunit gamma; Laminin-9 subunit beta; laminin-9 subunit gamma; LAMS; NPHS5; npht; SLAM; S-LAM beta; S-LAM gamma; S-laminin subunit beta; S-laminin subunit gamma | |
Mouse | |
Protein A | |
RUO | |
3913, 3915 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction