Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin beta-2/gamma-1 Monoclonal Antibody (M046)
GREENER_CHOICE

Catalog No. PIMA547722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547722 Supplier Invitrogen™ Supplier No. MA547722
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone M046 detects native laminin isoforms that include both laminin beta2 and gamma1 subunits. In native immunoblots, the antibody detects laminin 521, laminin 221, and laminin 121, but does not detect lamin 332 and laminin 411. The antibody weakly detects laminin 511. This antibody can be used for immunocytochemical detection of laminin beta2/gamma1 subunits, and can also be used for detection of native laminin isoforms in immunoblots and ELISA.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin beta-2/gamma-1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone M046
Concentration ~1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene LAMC1
Gene Accession No. P11047, P55268
Gene Alias AA408497; AW211941; B2e; basement membrane protein; Lama; Lamb2; Lamb-2; Lamc1; Laminin B1s chain; laminin B2 chain; laminin beta 2; laminin beta 2 chain; laminin chain B3; laminin chain beta 2; laminin gamma 1 chain; Laminin S; laminin subunit beta 2; laminin subunit beta-2; laminin subunit gamma 1; laminin subunit gamma-1; laminin, beta 2; laminin, beta 2 (laminin S); laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1 subunit gamma; Laminin-10 subunit gamma; laminin-11 subunit beta; Laminin-11 subunit gamma; laminin-14 subunit beta; Laminin-15 subunit beta; laminin-2 subunit gamma; Laminin-3 subunit beta; laminin-3 subunit gamma; Laminin-4 subunit beta; laminin-4 subunit gamma; Laminin-6 subunit gamma; laminin-7 subunit beta; Laminin-7 subunit gamma; laminin-8 subunit gamma; Laminin-9 subunit beta; laminin-9 subunit gamma; LAMS; NPHS5; npht; SLAM; S-LAM beta; S-LAM gamma; S-laminin subunit beta; S-laminin subunit gamma
Gene Symbols LAMB2, LAMC1
Host Species Mouse
Immunogen Clone (M046) was generated from a recombinant human laminin 521 protein that includes a heterotrimer of three subunits laminin alpha5, laminin beta2, and laminin gamma1 subunits.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3913, 3915
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.