Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin beta-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103211 Supplier Thermo Scientific Supplier No. PA5103211
Add to Cart
Add to Cart

Laminin beta-2/gamma-1 Polyclonal Antibody

Laminin beta-2/gamma-1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin beta-2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LAMB2
Gene Accession No. P15800, P55268, Q61292
Gene Alias AW211941; Lamb2; Lamb-2; Laminin B1s chain; laminin chain B3; laminin chain beta 2; Laminin S; laminin subunit beta 2; laminin subunit beta-2; laminin, beta 2; laminin, beta 2 (laminin S); laminin-11 subunit beta; Laminin-14 subunit beta; laminin-15 subunit beta; Laminin-3 subunit beta; laminin-4 subunit beta; Laminin-7 subunit beta; laminin-9 subunit beta; LAMS; NPHS5; npht; SLAM; S-LAM beta; S-laminin subunit beta
Gene Symbols LAMB2
Host Species Rabbit
Immunogen A synthesized peptide derived from human LAMB2(Accession P55268), corresponding to amino acid residues G72-A122.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16779, 25473, 3913
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.