Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin beta 3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32122125UL
Description
Laminin beta 3 Polyclonal antibody specifically detects Laminin beta 3 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Laminin beta 3 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL | |
BM600-125KDA, Epiligrin subunit bata, FLJ99565, Kalinin B1 chain, Kalinin subunit beta, kalinin-140kDa, LAM5, Laminin 5, Laminin B1k chain, laminin S B3 chain, laminin subunit beta-3, laminin, beta 3, laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)), Laminin-5 subunit beta, LAMNB1, Nicein subunit beta, nicein-125kDa | |
This antibody was developed against Recombinant Protein corresponding to amino acids: KLVTSMTKQLGDFWTRMEELRHQARQQGAEAVQAQQLAEGASEQALSAQEGFERIKQKYAELKDRLGQSSMLGEQGARIQSVKTEAEELFGETMEMMDRMKDMELELLRGSQAIMLRSADLTGLEKRVEQ | |
25 μL | |
Primary | |
Human | |
Purified |
Immunocytochemistry | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
3914 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction