Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Laminin beta-3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552510
Description
Immunogen sequence: KLVTSMTKQL GDFWTRMEEL RHQARQQGAE AVQAQQLAEG ASEQALSAQE GFERIKQKYA ELKDRLGQSS MLGEQGARIQ SVKTEAEELF GETMEMMDRM KDMELELLRG SQAIMLRSAD LTGLEKRVEQ Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 76%.
The product encoded by this gene is a laminin that belongs to a family of basement membrane proteins. This protein is a beta subunit laminin, which together with an alpha and a gamma subunit, forms laminin-5. Mutations in this gene cause epidermolysis bullosa junctional Herlitz type, and generalized atrophic benign epidermolysis bullosa, diseases that are characterized by blistering of the skin. Multiple alternatively spliced transcript variants that encode the same protein have been found for this gene.
Specifications
Laminin beta-3 | |
Polyclonal | |
Unconjugated | |
LAMB3 | |
AI1A; BM600-125KDA; Epiligrin subunit bata; kalinin B1 chain; Kalinin subunit beta; kalinin-140kDa; LAM5; Lamb3; Laminin B1k chain; laminin beta 3; laminin chain; laminin S B3 chain; laminin subunit beta 3; laminin subunit beta-3; laminin, beta 3; laminin, beta 3 (nicein (125kD), kalinin (140kD), BM600 (125kD)); laminin-5 subunit beta; LAMNB1; Nicein subunit beta; nicein, 125kD; nicein, 125kDa; nicein-125k; nicein-125kDa; RP1-272L16.3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3914 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13751 | |
LAMB3 | |
Recombinant protein corresponding to Human Laminin beta-3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction