Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin gamma-1,2,3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579579
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579579 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579579 Supplier Invitrogen™ Supplier No. PA579579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody reacts will all 3 isoforms of Laminin gamma: Laminin gamma-1, gamma-2 and gamma-3. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A431 whole cell, human A549 whole cell, human PC-3 whole cell, human Caco-2 whole cellrat NRK whole cell, rat RH35 whole cell, mouse NIH/3T3 whole cell, mouse Hepa1-6 whole cell. IHC: human lung cancer tissue IHC-F: human placenta tissue.

Extracellular matrix (ECM) surrounds and separates most tissue compartments, determines tissue architecture and dynamically interacts with its surrounding cells. The basement membrane is a part of the ECM and is composed of collagens, proteoglycans, elastin and several glycoproteins of which laminin is the most abundant. Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains Dystroglycan is a major laminin receptor in muscle and epithelia, and nidogen is a high-affinity laminin-binding protein important for basement-membrane assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin gamma-1,2,3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation Antibody with 5mg BSA, 0.2mg Na2PO4, 0.9mg NaCl and 0.05mg sodium azide
Gene LAMC1
Gene Accession No. P02468, P11047, Q13753, Q61092, Q9R0B6, Q9Y6N6
Gene Alias AA589349; AI562206; alternatively spliced laminin gamma2 chain; AW240805; B2e; B2T; BM600; BM600-100kDa; cell-scattering factor 140 kDa subunit; CSF; CSF 140 kDa subunit; EBR2; EBR2A; epiligrin subunit gamma; Kalinin subunit gamma; kalinin/nicein/epiligrin 100 kDa subunit; Ladsin 140 kDa subunit; LAMB2; LAMB2T; LAMC1; Lamc2; Lamc3; lamimin, gamma 2; laminin B2 chain; Laminin B2t chain; laminin gamma 3; laminin gamma2 chain; laminin subunit gamma 1; laminin subunit gamma 2; laminin subunit gamma 3; laminin subunit gamma-1; Laminin subunit gamma-2; laminin subunit gamma-3; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin, gamma 2; laminin, gamma 3; laminin-1 subunit gamma; Laminin-10 subunit gamma; laminin-11 subunit gamma; Laminin-12 subunit gamma; laminin-14 subunit gamma; Laminin-15 subunit gamma; laminin-2 subunit gamma; Laminin-3 subunit gamma; laminin-4 subunit gamma; Laminin-5 subunit gamma; laminin-6 subunit gamma; Laminin-7 subunit gamma; laminin-8 subunit gamma; Laminin-9 subunit gamma; LAMNB2; Large adhesive scatter factor 140 kDa subunit; nicein subunit gamma; nicein, 100kD; nicein, 100kDa; OCCM; S-LAM gamma; S-laminin subunit gamma
Gene Symbols LAMC1, LAMC2, LAMC3
Host Species Rabbit
Immunogen Peptide mixture of laminin gamma1,2,3 (DLNKLNEIEGTLNKAKDEMK; MSELEERARQQRGHLHLLET; LQRKLSLLEQESQQQELQIQ).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10319, 117036, 16782, 192362, 226519, 23928, 311862, 3915, 3918
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.