Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin gamma 1 Antibody (CL3195) - Azide and BSA Free, Novus Biologicals Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP344406
Description
Laminin gamma 1 Monoclonal antibody specifically detects Laminin gamma 1 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)Specifications
Laminin gamma 1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
LAMB2Laminin-7 subunit gamma, Laminin B2 chain, laminin subunit gamma-1, laminin, gamma 1 (formerly LAMB2), Laminin-1 subunit gamma, Laminin-10 subunit gamma, Laminin-11 subunit gamma, Laminin-2 subunit gamma, Laminin-3 subunit gamma, Laminin-4 subunit gamma, Laminin-6 subunit gamma, Laminin-8 subunit gamma, Laminin-9 subunit gamma, MGC87297, S-LAM gamma, S-laminin subunit gamma | |
This antibody was generated using a recombinant protein sequence of P11047, with the exact immunogen sequence remaining proprietary. | |
100 μg | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Tumor Suppressors | |
3915 | |
Human | |
Antibody | |
IgG2b |
Western Blot, Immunohistochemistry (Paraffin) | |
CL3195 | |
Unconjugated | |
Lyophilized from a 0.2 um filtered solution in PBS with Trehalose | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
Store at 4°C short term. Aliquot and store at -24°C long term. Avoid freeze-thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction