Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin gamma 1 Antibody (SR1311), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Laminin gamma 1 |
---|---|
Clone | SR1311 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
Laminin gamma 1 Monoclonal antibody specifically detects Laminin gamma 1 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
Laminin gamma 1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
LAMB2Laminin-7 subunit gamma, Laminin B2 chain, laminin subunit gamma-1, laminin, gamma 1 (formerly LAMB2), Laminin-1 subunit gamma, Laminin-10 subunit gamma, Laminin-11 subunit gamma, Laminin-2 subunit gamma, Laminin-3 subunit gamma, Laminin-4 subunit gamma, Laminin-6 subunit gamma, Laminin-8 subunit gamma, Laminin-9 subunit gamma, MGC87297, S-LAM gamma, S-laminin subunit gamma | |
A synthesized peptide derived from human Laminin gamma 1 (Uniprot #: P11047, Q5VYE7, Q6NVY8) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1311 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3915 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title