Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin gamma-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536300 Supplier Invitrogen™ Supplier No. PA536300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 178 kDa. Purity is >95% by SDS-PAGE.

Laminins are essential and abundant structural non-collagenous glycoproteins localizing to basement membranes. Basement membranes (cell-associated extracellular matrices (ECMs)) are polymers of Laminins with stabilizing Type IV Collagen networks, Nidogen and several proteoglycans. Basement membranes are found under epithelial layers, around the endothelium of blood vessels, and surrounding muscle, peripheral nerve and fat cells. Formation of basement membranes influences cell proliferation, phenotype, migration, gene expression and tissue architecture. Each Laminin is a heterotrimer of alpha, beta and gamma chain subunits that undergoes cell-secretion and incorporation into the ECM. Laminins can self-assemble, bind to other matrix macromolecules and have unique and shared cell interactions mediated by Integrins, dystroglycan and cognate Laminin receptors. The human Laminin gamma-1 gene maps to chromosome 1q31 and is ubiquitously expressed in tissues that produce basement membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin gamma-1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene LAMC1
Gene Accession No. P11047
Gene Alias AA408497; B2e; basement membrane protein; LAMA; LAMA1; Lama-1; LAMB2; LAMC1; Laminin A chain; laminin B2 chain; laminin gamma 1 chain; laminin S; laminin subunit alpha 1; laminin subunit alpha-1; laminin subunit gamma 1; laminin subunit gamma-1; laminin, alpha 1; laminin, beta 2; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1; laminin-1 subunit alpha; Laminin-1 subunit gamma; laminin-10 subunit gamma; Laminin-11 subunit gamma; laminin-2 subunit gamma; Laminin-3 subunit alpha; laminin-3 subunit gamma; Laminin-4 subunit gamma; laminin-6 subunit gamma; Laminin-7 subunit gamma; laminin-8 subunit gamma; Laminin-9 subunit gamma; PTBHS; S-LAM alpha; S-LAM gamma; S-LAM-alpha; S-laminin subunit alpha; S-laminin subunit gamma
Gene Symbols LAMC1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 1441-1490 of human Laminin gamma-1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3915
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.