Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin gamma-2 Monoclonal Antibody (CL2980)
GREENER_CHOICE

Catalog No. PIMA524646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524646 Supplier Invitrogen™ Supplier No. MA524646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: NAGVTIQDTL NTLDGLLHLM DQPLSVDEEG LVLLEQKLSR AKTQINSQLR PMMSELEERA RQQRGHLHLL ETSIDGILAD VKNLEN Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 79%. Binds to an epitope located within the peptide sequence IQDTLNTLDGLLHLM as determined by overlapping synthetic peptides.

Laminins, a family of extracellular matrix glycoproteins, are the major noncollagenous constituent of basement membranes. They have been implicated in a wide variety of biological processes including cell adhesion, differentiation, migration, signaling, neurite outgrowth and metastasis. Laminins are composed of 3 non identical chains: laminin alpha, beta and gamma (formerly A, B1, and B2, respectively) and they form a cruciform structure consisting of 3 short arms, each formed by a different chain, and a long arm composed of all 3 chains. Each laminin chain is a multidomain protein encoded by a distinct gene. Several isoforms of each chain have been described. Different alpha, beta and gamma chain isomers combine to give rise to different heterotrimeric laminin isoforms which are designated by Arabic numerals in the order of their discovery, i.e. alpha1beta1gamma1 heterotrimer is laminin 1. The biological functions of the different chains and trimer molecules are largely unknown, but some of the chains have been shown to differ with respect to their tissue distribution, presumably reflecting diverse functions in vivo. This gene encodes the gamma chain isoform laminin, gamma 2. The gamma 2 chain, formerly thought to be a truncated version of beta chain (B2t), is highly homologous to the gamma 1 chain; however, it lacks domain VI, and domains V, IV and III are shorter. It is expressed in several fetal tissues but differently from gamma 1, and is specifically localized to epithelial cells in skin, lung and kidney. The gamma 2 chain together with alpha 3 and beta 3 chains constitute laminin 5 (earlier known as kalinin), which is an integral part of the anchoring filaments that connect epithelial cells to the underlying basement membrane. The epithelium-specific expression of the gamma 2 chain implied its role as an epithelium attachment molecule, and mutations in this gene have been associated with junctional epidermolysis bullosa, a skin disease characterized by blisters due to disruption of the epidermal-dermal junction. Two transcript variants resulting from alternative splicing of the 3' terminal exon, and encoding different isoforms of gamma 2 chain, have been described. The two variants are differentially expressed in embryonic tissues, however, the biological significance of the two forms is not known. Transcript variants utilizing alternative polyA_signal have also been noted in literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin gamma-2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CL2980
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Lamc2
Gene Accession No. Q13753
Gene Alias AA589349; alternatively spliced laminin gamma2 chain; B2T; BM600; BM600-100kDa; cell-scattering factor 140 kDa subunit; CSF; CSF 140 kDa subunit; EBR2; EBR2A; Epiligrin subunit gamma; kalinin subunit gamma; Kalinin/nicein/epiligrin 100 kDa subunit; ladsin 140 kDa subunit; LAM LAMNB2; LAMB2T; LAMC2; lamimin, gamma 2; laminin B2t chain; laminin gamma2 chain; laminin subunit gamma 2; laminin subunit gamma-2; laminin, gamma 2; laminin-5 subunit gamma; LAMNB2; Large adhesive scatter factor 140 kDa subunit; nicein subunit gamma; nicein, 100kD; nicein, 100kDa
Gene Symbols Lamc2
Host Species Mouse
Immunogen Recombinant protein fragment of Human LAMC2 (Product #RP-93997)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3918
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.