Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin Monoclonal Antibody (ABM412), Abbomax

Catalog No. 502203812
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3812 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3812 Supplier Abbomax Inc Supplier No. 5007794
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Lung, heart Cellular location: Extracellular matrix, Cytoplasmic.

Laminin Monoclonal antibody specifically detects Laminin in Bovine, Chicken, Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Laminin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ABM412
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene LAMC1
Gene Accession No. P07942, P11047, P24043, P25391, Q01635
Gene Alias 5830440B04; AA408497; B2e; basement membrane protein; C77966; C80098; C81607; CLM; D130003D08Rik; dy; dystrophia muscularis; LAMA; LAMA1; Lama-1; Lama2; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; Lamb2; LAMC1; Laminin A chain; laminin alpha 2 subunit; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin B2 chain; laminin beta-1 chain; laminin gamma 1 chain; laminin M chain; laminin S; laminin subunit alpha 1; laminin subunit alpha 2; laminin subunit alpha-1; Laminin subunit alpha-2; laminin subunit beta 1; laminin subunit beta-1; laminin subunit gamma 1; laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, beta 2; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1; laminin-1 subunit alpha; Laminin-1 subunit beta; laminin-1 subunit gamma; Laminin-10 subunit beta; laminin-10 subunit gamma; Laminin-11 subunit gamma; laminin-12 subunit alpha; Laminin-12 subunit beta; laminin-2 subunit alpha; Laminin-2 subunit beta; laminin-2 subunit gamma; Laminin-3 subunit alpha; laminin-3 subunit gamma; Laminin-4 subunit alpha; laminin-4 subunit gamma; Laminin-6 subunit beta; laminin-6 subunit gamma; Laminin-7 subunit gamma; laminin-8 subunit beta; Laminin-8 subunit gamma; laminin-9 subunit gamma; LAMM; LIS5; LOW QUALITY PROTEIN: laminin subunit alpha-1; LOW QUALITY PROTEIN: laminin subunit alpha-2; LOW QUALITY PROTEIN: laminin subunit beta-1; mer; merosin; merosin heavy chain; mKIAA4087; PTBHS; S-LAM alpha; S-LAM gamma; S-LAM-alpha; S-laminin subunit alpha; S-laminin subunit gamma
Host Species Mouse
Immunogen Purified recombinant partial length of human Laminin protein
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100138434, 117036, 284217, 298941, 309368, 316758, 374016, 3908, 3912, 3915, 396478, 421709, 424442, 506387, 520030, 532572
Target Species Bovine, Chicken, Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.