Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin Monoclonal Antibody (LN82-13), QED Bioscience™
SDP

Catalog No. 502278033
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8033 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8033 Supplier Leinco Technologies Inc Supplier No. 42035100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Laminin Monoclonal antibody specifically detects Laminin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Laminin
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LN82-13
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and no preservative; pH 7.4
Gene LAMC1
Gene Accession No. P07942, P11047, P24043, P25391
Gene Alias 5830440B04; AA408497; B2e; basement membrane protein; C77966; C80098; C81607; CLM; D130003D08Rik; dy; dystrophia muscularis; LAMA; LAMA1; Lama-1; Lama2; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; Lamb2; LAMC1; Laminin A chain; laminin alpha 2 subunit; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin B2 chain; laminin beta-1 chain; laminin gamma 1 chain; laminin M chain; laminin S; laminin subunit alpha 1; laminin subunit alpha 2; laminin subunit alpha-1; Laminin subunit alpha-2; laminin subunit beta 1; laminin subunit beta-1; laminin subunit gamma 1; laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, beta 2; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1; laminin-1 subunit alpha; Laminin-1 subunit beta; laminin-1 subunit gamma; Laminin-10 subunit beta; laminin-10 subunit gamma; Laminin-11 subunit gamma; laminin-12 subunit alpha; Laminin-12 subunit beta; laminin-2 subunit alpha; Laminin-2 subunit beta; laminin-2 subunit gamma; Laminin-3 subunit alpha; laminin-3 subunit gamma; Laminin-4 subunit alpha; laminin-4 subunit gamma; Laminin-6 subunit beta; laminin-6 subunit gamma; Laminin-7 subunit gamma; laminin-8 subunit beta; Laminin-8 subunit gamma; laminin-9 subunit gamma; LAMM; LIS5; LOW QUALITY PROTEIN: laminin subunit alpha-1; LOW QUALITY PROTEIN: laminin subunit alpha-2; LOW QUALITY PROTEIN: laminin subunit beta-1; mer; merosin; merosin heavy chain; mKIAA4087; PTBHS; S-LAM alpha; S-LAM gamma; S-LAM-alpha; S-laminin subunit alpha; S-laminin subunit gamma
Host Species Mouse
Immunogen Human laminin.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284217, 3908, 3912, 3915
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.