Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Laminin Polyclonal Antibody, DyLight™ 550
GREENER_CHOICE

Catalog No. PIPA522903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522903 Supplier Invitrogen™ Supplier No. PA522903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is pan-specific and reacts well with all Laminin isoforms tested: Laminin-1 (alpha-1, beta-1, and gamma-1) and Laminin-2 (alpha-2, beta-1, and gamma-1). The antibody functionally inhibits Laminin in mouse and rat. It binds to Laminin and inhibits most, if not all, of its cell adhesion and growth promotive properties. Immunostaining is enhanced by antigen retrieval with pepsin, especially paraffin tissue. Suggested positive control: basement membranes (blood vessels of any tissue, tumors, nerve).

Laminin is a crucial glycoprotein that plays a significant role in the extracellular matrix, particularly within the basement membrane. It is composed of three subunits: alpha, beta, and gamma chains. These chains are disulfide-linked polypeptides that together form a heterotrimer, resembling the shape of a cross. The human genome encodes 11 genetically distinct laminin chains, which combine to form over 14 different laminin isoforms, each with specific tissue distributions and functions. Laminins can bind to other laminin molecules, as well as integrins and other matrix proteins, providing structural support and influencing cell differentiation, migration, and adhesion. These glycoproteins are vital during early embryonic development, where they contribute to the formation of basement membrane-like structures, even in the absence of type IV collagen. The evolutionary origin of laminin gene families can be traced back to early branching animals, providing insights into metazoan gene family development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Laminin
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.70 mg/mL
Conjugate DyLight 550
Formulation 50mM sodium borate with 0.05% sodium azide
Gene LAMC1
Gene Accession No. P02468, P02469, P07942, P11047, P19137, P24043, P25391, Q60675
Gene Alias 5830440B04; AA408497; B2e; basement membrane protein; C77966; C80098; C81607; CLM; D130003D08Rik; dy; dystrophia muscularis; LAMA; LAMA1; Lama-1; Lama2; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; Lamb2; LAMC1; Laminin A chain; laminin alpha 2 subunit; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin B2 chain; laminin beta-1 chain; laminin gamma 1 chain; laminin M chain; laminin S; laminin subunit alpha 1; laminin subunit alpha 2; laminin subunit alpha-1; Laminin subunit alpha-2; laminin subunit beta 1; laminin subunit beta-1; laminin subunit gamma 1; laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, beta 2; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1; laminin-1 subunit alpha; Laminin-1 subunit beta; laminin-1 subunit gamma; Laminin-10 subunit beta; laminin-10 subunit gamma; Laminin-11 subunit gamma; laminin-12 subunit alpha; Laminin-12 subunit beta; laminin-2 subunit alpha; Laminin-2 subunit beta; laminin-2 subunit gamma; Laminin-3 subunit alpha; laminin-3 subunit gamma; Laminin-4 subunit alpha; laminin-4 subunit gamma; Laminin-6 subunit beta; laminin-6 subunit gamma; Laminin-7 subunit gamma; laminin-8 subunit beta; Laminin-8 subunit gamma; laminin-9 subunit gamma; LAMM; LIS5; LOW QUALITY PROTEIN: laminin subunit alpha-1; LOW QUALITY PROTEIN: laminin subunit alpha-2; LOW QUALITY PROTEIN: laminin subunit beta-1; mer; merosin; merosin heavy chain; mKIAA4087; PTBHS; S-LAM alpha; S-LAM gamma; S-LAM-alpha; S-laminin subunit alpha; S-laminin subunit gamma
Gene Symbols LAMA1, LAMA2, LAMB1, LAMC1
Host Species Rabbit
Immunogen Laminin 111 isolated from mouse Engelbreth-Holm-Swarm (EHS) sarcoma cells.
Purification Method IgG fraction
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100350500, 100357482, 100358679, 100757343, 100761564, 100765555, 101102692, 101105588, 101105743, 101117424, 103346447, 117036, 16772, 16773, 16777, 226519, 284217, 298941, 309368, 316758, 3908, 3912, 3915
Target Species Human, Mouse, Rat, Rabbit, Sheep, Hamster, Mammalia, Invertebrate
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.