Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin Rabbit anti-Human, Biotin, Polyclonal, Rockland™
SDP

Catalog No. 501992443
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2443 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2443 Supplier Rockland Immunochemicals Supplier No. 600406116
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. Restore with 1.0 mL of deionized water (or equivalent). For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Typically less than 1% cross-reactivity against other extracellular matrix proteins was detected by ELISA against purified standards. This antibody reacts with most mammalian Laminins and has negligible cross-reactivity with Type I, II, III, IV, V or VI Collagens or Fibronectin. Non-specific cross-reaction of anti-Laminin antibodies with other human serum proteins or non-Laminin extracellular matrix proteins is negligible.

Laminin antibodies are widely used to label blood vessels and basement membranes.

Specifications

Antigen Laminin
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation 0.02M potassium phosphate with 0.15M NaCl, 10 mg/mL BSA and 0.01% sodium azide; pH 7.2
Gene LAMA2
Gene Accession No. P07942, P11047, P24043, P25391
Gene Alias 5830440B04; AA408497; B2e; C77966; C80098; C81607; CLM; D130003D08Rik; dy; dystrophia muscularis; LAMA; Lama1; Lama-1; Lama2; LAMB1; Lamb-1; Lamb1-1; LAMB2; LAMC1; Laminin A chain; laminin alpha 2 subunit; laminin B1; Laminin B1 chain; laminin B1 subunit 1; Laminin B2 chain; Laminin M chain; laminin subunit alpha 1; laminin subunit alpha 2; Laminin subunit alpha-1; Laminin subunit alpha-2; laminin subunit beta 1; Laminin subunit beta-1; laminin subunit gamma 1; Laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1; Laminin-1 subunit alpha; Laminin-1 subunit beta; Laminin-1 subunit gamma; Laminin-10 subunit beta; Laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit alpha; Laminin-12 subunit beta; Laminin-2 subunit alpha; Laminin-2 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit alpha; Laminin-3 subunit gamma; Laminin-4 subunit alpha; Laminin-4 subunit gamma; Laminin-6 subunit beta; Laminin-6 subunit gamma; Laminin-7 subunit gamma; Laminin-8 subunit beta; Laminin-8 subunit gamma; Laminin-9 subunit gamma; LAMM; LIS5; mer; merosin; Merosin heavy chain; mKIAA4087; PTBHS; S-LAM alpha; S-LAM gamma; S-LAM-alpha; S-laminin subunit alpha; S-laminin subunit gamma
Gene Symbols LAMA1, LAMA2, LAMB1, LAMC1
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284217, 3908, 3912, 3915
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.