Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

laminin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980534
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0534 Supplier Bioss Supplier No. BS0821R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Laminin antibodies are widely used to label blood vessels and basement membranes.

Specifications

Antigen laminin
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene LAMA2
Gene Accession No. P07942, P11047, P24043, P25391
Gene Alias 5830440B04; AA408497; B2e; C77966; C80098; C81607; CLM; D130003D08Rik; dy; dystrophia muscularis; LAMA; Lama1; Lama-1; Lama2; LAMB1; Lamb-1; Lamb1-1; LAMB2; LAMC1; Laminin A chain; laminin alpha 2 subunit; laminin B1; Laminin B1 chain; laminin B1 subunit 1; Laminin B2 chain; Laminin M chain; laminin subunit alpha 1; laminin subunit alpha 2; Laminin subunit alpha-1; Laminin subunit alpha-2; laminin subunit beta 1; Laminin subunit beta-1; laminin subunit gamma 1; Laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1; Laminin-1 subunit alpha; Laminin-1 subunit beta; Laminin-1 subunit gamma; Laminin-10 subunit beta; Laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit alpha; Laminin-12 subunit beta; Laminin-2 subunit alpha; Laminin-2 subunit beta; Laminin-2 subunit gamma; Laminin-3 subunit alpha; Laminin-3 subunit gamma; Laminin-4 subunit alpha; Laminin-4 subunit gamma; Laminin-6 subunit beta; Laminin-6 subunit gamma; Laminin-7 subunit gamma; Laminin-8 subunit beta; Laminin-8 subunit gamma; Laminin-9 subunit gamma; LAMM; LIS5; mer; merosin; Merosin heavy chain; mKIAA4087; PTBHS; S-LAM alpha; S-LAM gamma; S-LAM-alpha; S-laminin subunit alpha; S-laminin subunit gamma
Gene Symbols LAMA1, LAMA2, LAMB1, LAMC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Laminin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284217, 3908, 3912, 3915
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.