Learn More
Laminin Rat anti-Human, Mouse, Porcine, Clone: A5, MUbio

Rat Monoclonal Antibody
Supplier: Vector Laboratories MUB1100P

Description
RNF402 reacts with both the phosphorylated and non-phosphorylated isoform of the 200 kD neurofilament protein. The antibody may be stored at 4 C. For prolonged storage prepare appropriate aliquots and store at or below -20 C. Prior to use, an aliquot is thawed slowly in the dark at ambient temperature, spun down again and used to prepare working dilutions by adding sterile phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. Working dilutions should be stored at 4 C, not refrozen, and preferably used the same day. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance or the concentration of the product.
Laminin Monoclonal antibody specifically detects Laminin in Human, Mouse, Porcine samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)Specifications
Laminin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH | |
P02468, P02469, P07942, P11047, P19137, P24043, P25391, Q60675 | |
Rat | |
Protein A/G | |
RUO | |
100154420, 100514785, 100625589, 16772, 16773, 16777, 226519, 284217, 3908, 3912, 3915, 396707 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
A5 | |
Unconjugated | |
LAMC1 | |
5830440B04; AA408497; B2e; C77966; C80098; C81607; CLM; D130003D08Rik; dy; dystrophia muscularis; LAMA; Lama1; Lama-1; Lama2; LAMB1; Lamb-1; Lamb1-1; Lamb2; LAMC1; laminin A chain; laminin alpha 2 subunit; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin B2 chain; Laminin M chain; laminin subunit alpha 1; laminin subunit alpha 2; laminin subunit alpha-1; Laminin subunit alpha-2; laminin subunit beta 1; laminin subunit beta-1; laminin subunit gamma 1; laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1; laminin-1 subunit alpha; Laminin-1 subunit beta; laminin-1 subunit gamma; Laminin-10 subunit beta; laminin-10 subunit gamma; Laminin-11 subunit gamma; Laminin-12 subunit alpha; Laminin-12 subunit beta; laminin-2 subunit alpha; Laminin-2 subunit beta; laminin-2 subunit gamma; Laminin-3 subunit alpha; laminin-3 subunit gamma; Laminin-4 subunit alpha; laminin-4 subunit gamma; Laminin-6 subunit beta; laminin-6 subunit gamma; Laminin-7 subunit gamma; laminin-8 subunit beta; Laminin-8 subunit gamma; laminin-9 subunit gamma; LAMM; LIS5; mer; merosin; Merosin heavy chain; mKIAA4087; PTBHS; S-LAM alpha; S-LAM gamma; S-LAM-alpha; S-laminin subunit alpha; S-laminin subunit gamma | |
Laminin | |
0.1 mg | |
Primary | |
Human, Mouse, Pig | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.