Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin S/Laminin beta 2 Antibody (CL2979) - Azide and BSA Free, Novus Biologicals Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP344401
Description
Laminin S/Laminin beta 2 Monoclonal antibody specifically detects Laminin S/Laminin beta 2 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)Specifications
Laminin S/Laminin beta 2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Laminin B1s chain, laminin S, laminin subunit beta-2, laminin, beta 2 (laminin S), Laminin-11 subunit beta, Laminin-14 subunit beta, Laminin-15 subunit beta, Laminin-3 subunit beta, Laminin-4 subunit beta, Laminin-7 subunit beta, LAMSLaminin-9 subunit beta, S-LAM beta, S-laminin subunit beta | |
This antibody was generated using a recombinant protein sequence of P55268, with the exact immunogen sequence remaining proprietary. | |
100 μg | |
Primary | |
Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL. | |
Store at 4°C short term. Aliquot and store at -24°C long term. Avoid freeze-thaw cycles. | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
CL2979 | |
Unconjugated | |
Lyophilized from a 0.2 um filtered solution in PBS with Trehalose | |
Mouse | |
Protein A purified | |
RUO | |
3913 | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction