Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin S/Laminin beta 2 Antibody (OTI3B4), DyLight 680, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP271114FR

Catalog No. NBP271114FR


Only null left
Add to Cart

Description

Description

Laminin S/Laminin beta 2 Monoclonal specifically detects Laminin S/Laminin beta 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

Laminin S/Laminin beta 2
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
LAMB2
Recombinant protein fragment corresponding to amino acids 314-561 of human Laminin S/Laminin beta 2 (NP_002283) produced in E.coli.
0.1 mL
3913.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI3B4
Western Blot, Immunohistochemistry
Laminin B1s chain, laminin S, laminin subunit beta-2, laminin, beta 2 (laminin S), Laminin-11 subunit beta, Laminin-14 subunit beta, Laminin-15 subunit beta, Laminin-3 subunit beta, Laminin-4 subunit beta, Laminin-7 subunit beta, LAMSLaminin-9 subunit beta, S-LAM beta, S-laminin subunit beta
Mouse
Affinity Purified
Primary
Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.