Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ SMART 3™ Colorimeter Reagent Systems



Click to view available options
Type:
Carbohydrazide Reagent
Copper Cuprizone Reagent
Fluoride Reagent
Hydrogen Peroxide LR Reagent
Lead Reagent
Molybdenum HR Reagent, Thioglycolate
Nitrogen Total Reagent
Phosphorus Total HR Reagent
Phosphorus Total LR Reagent
Surfactant Reagent
Tolyltriazole Reagent
Range:
0 to 0.9 ppm
0 to 1.5 ppm
0 to 2 ppm
0 to 3.5 ppm
0 to 5 ppm
0 to 8 ppm
0.0 to 50 ppm
0.0 to 70 ppm
1 to 30 ppm
3 to 25 ppm
No. of Tests:
25
50
100
Test Method:
Ascorbic acid or digestion
Bromophenol blue
Chromotropic acid or digestion
Cuprizone
DPD
Iron reduction
Molybdovanadate or Digestion
PAR
SPADNS
Thioglycolate
UV Oxidation or dichromate
Description
- Reagents for use in water testing and environmental studies.
Specifications
Specifications
For Use With (Equipment) | SMART 3 Colorimeter |
No. of Tests | 50 |
Range | 0 to 2 ppm |
Test Method | Cuprizone |
Type | Copper Cuprizone Reagent |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction