Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        LaMotte™ SMART 3™ Colorimeter Reagent Systems

    
    
    
        
            Shop All LaMotte Company Products
        
    
    
    
    
    
    
    
    
        

                    Click to view available options
                
                
                    
                            
                                Type:
                            
                        
                        
                            
                                Carbohydrazide Reagent
                            
                        
                            
                                Copper Cuprizone Reagent
                            
                        
                            
                                Fluoride Reagent
                            
                        
                            
                                Hydrogen Peroxide LR Reagent
                            
                        
                            
                                Lead Reagent
                            
                        
                            
                                Molybdenum HR Reagent, Thioglycolate
                            
                        
                            
                                Nitrogen Total Reagent
                            
                        
                            
                                Phosphorus Total HR Reagent
                            
                        
                            
                                Phosphorus Total LR Reagent
                            
                        
                            
                                Surfactant Reagent
                            
                        
                            
                                Tolyltriazole Reagent
                            
                        
                    
                            
                                Range:
                            
                        
                        
                            
                                0 to 0.9 ppm
                            
                        
                            
                                0 to 1.5 ppm
                            
                        
                            
                                0 to 2 ppm
                            
                        
                            
                                0 to 3.5 ppm
                            
                        
                            
                                0 to 5 ppm
                            
                        
                            
                                0 to 8 ppm
                            
                        
                            
                                0.0 to 50 ppm
                            
                        
                            
                                0.0 to 70 ppm
                            
                        
                            
                                1 to 30 ppm
                            
                        
                            
                                3 to 25 ppm
                            
                        
                    
                            
                                No. of Tests:
                            
                        
                        
                            
                                25
                            
                        
                            
                                50
                            
                        
                            
                                100
                            
                        
                    
                            
                                Test Method:
                            
                        
                        
                            
                                Ascorbic acid or digestion
                            
                        
                            
                                Bromophenol blue
                            
                        
                            
                                Chromotropic acid or digestion
                            
                        
                            
                                Cuprizone
                            
                        
                            
                                DPD
                            
                        
                            
                                Iron reduction
                            
                        
                            
                                Molybdovanadate or Digestion
                            
                        
                            
                                PAR
                            
                        
                            
                                SPADNS
                            
                        
                            
                                Thioglycolate
                            
                        
                            
                                UV Oxidation or dichromate
                            
                        
                    Description
- Reagents for use in water testing and environmental studies.
 
Specifications
Specifications
| For Use With (Equipment) | SMART 3 Colorimeter | 
| No. of Tests | 50 | 
| Range | 0.0 to 50 ppm | 
| Test Method | Thioglycolate | 
| Type | Molybdenum HR Reagent, Thioglycolate | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction