Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ SMART 3™ Colorimeter Reagent Systems
SDPSCI_ED

Catalog No. p-4237647 Shop All LaMotte Company Products
Click to view available options
Type:
Carbohydrazide Reagent
Copper Cuprizone Reagent
Fluoride Reagent
Hydrogen Peroxide LR Reagent
Lead Reagent
Molybdenum HR Reagent, Thioglycolate
Nitrogen Total Reagent
Phosphorus Total HR Reagent
Phosphorus Total LR Reagent
Surfactant Reagent
Tolyltriazole Reagent
Range:
0 to 0.9 ppm
0 to 1.5 ppm
0 to 2 ppm
0 to 3.5 ppm
0 to 5 ppm
0 to 8 ppm
0.0 to 50 ppm
0.0 to 70 ppm
1 to 30 ppm
3 to 25 ppm
No. of Tests:
25
50
100
Test Method:
Ascorbic acid or digestion
Bromophenol blue
Chromotropic acid or digestion
Cuprizone
DPD
Iron reduction
Molybdovanadate or Digestion
PAR
SPADNS
Thioglycolate
UV Oxidation or dichromate

Catalog No. S95011

Supplier: LaMotte™ 403101

Only null left
Add to Cart
Add to Cart

SMART 3™ Colorimeter Reagent Systems for use in water quality testing and environmental studies.

  • Reagents for use in water testing and environmental studies.

Specifications

For Use With (Equipment) SMART 3 Colorimeter
No. of Tests 50
Range 0 to 5 ppm
Test Method PAR
Type Lead Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.