Description
- Reagents for use in water testing and environmental studies.
Specifications
Specifications
| For Use With (Equipment) | SMART 3 Colorimeter |
| No. of Tests | 50 |
| Range | 0 to 8 ppm |
| Test Method | Bromophenol blue |
| Type | Surfactant Reagent |