Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Chlorine Individual Test Kit
SDPSCI_ED

Catalog No. s94976 Shop All LaMotte Company Products
Click to view available options
Quantity:
50 Tests

Catalog No. S94976

Supplier: LaMotte™ 317602

Only null left
Add to Cart
Add to Cart

Includes

  • Control reagent
  • DPD #1 powder
  • Ferrous Aluminum Sulfate
  • Chlorine DPD #3R blister tabs

Direct reading titrator method results in reliable, accurate reading

Uses advanced DPD-FAS titration to pinpoint chlorine levels — free available chlorine, total chlorine and combined chlorine

  • Range/sensitivity: 0 to 10ppm Cl in 0.2ppm increments
  • Number of tests: 50 at 10ppm
  • Number of reagents per test: 4
  • DPD = N, N-Diethyl-1, 4-phenylene-diamine sulfate, FAS = Ferrous aluminum sulfate, PPM = Parts Per Million, Cl = Chlorine.
  • Ideal for testing chlorine levels in drinking water, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater

Specifications

For Use With (Application) Ideal for testing chlorine levels in drinking water, food and beverage, industrial water boiling and cooling, laundry and sanitation, water and wastewater
Includes Control reagent, DPD 1 powder, ferrous aluminum sulfate, chlorine DPD 3R blister tabs
Quantity 50 Tests
Range 0 to 10 ppm
Test Method Direct-reading titrator
Type Chlorine Test Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.