Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips



Click to view available options
Parameter Specifications:
Chlorine - total, low range
Hydrogen Peroxide
Nitrate and Nitrite
Peracetic acid
Quantity:
25 Strips/Vial
50 Strips/Vial
Range:
0 to 10 ppm
0 to 50 ppm
0-10-20-40-60-85-160 ppm
0-5-10-25-50, 0-0.5-1-5-10
Description
- No reagents or field kits required for most strips
- Chemically treated pads on plastic strips produce color change
- Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
- As little as 10-second development time
Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †
†Strips have been evaluated for use in these applications. Use in other applications is subject to potential interferences.
Specifications
Specifications
For Use With (Application) | Water quality testing |
Parameter Specifications | Hydrogen Peroxide |
Quantity | 25 Strips/Vial |
Range | 0 to 50 ppm |
Storage Requirements | Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight |
Type | Single Factor Test Strip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction